Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-(+)-3-Hydroxy-3-phenylpropionic acid
(R)-(+)-3-Hydroxy-3-phenylpropionic acid ≥98%
(R)-(+)-3-Hydroxy-3-phenylpropionic acid ≥98%
Irritant
(R)-(+)-3-Hydroxy-3-phenylpropionic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
(R)-(+)-3-Hydroxy-3-phenylpropionic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
2768-42-5
:
Formula:
C₉H₁₀O₃
MDL Number:
MFCD00145219
:
166.18 g/mol
Melting Pt:
118…121 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL19040
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation