Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Heptyltin trichloride
Heptyltin trichloride
Heptyltin trichloride
Heptyltin trichloride
Catalog # PROODE-MS-03441
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Certificates
Expand image
Heptyltin trichloride
Catalog # PROODE-MS-03441
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Certificates
Specifications
Pack type:
Glass bottle for solids
Pk:
300 mg
Seq:
2625
Specifications
About this item
Product Details
Documentation
:
CAS Number:
59344-47-7
Formula:
C₇H₁₅Cl₃Sn
Product Details
Documentation