(Dichloromethyl)dimethylchlorosilane
Catalog # ABCRAB111035.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
:
EINECS:242-058-1
CAS Number:18171-59-0
Formula:C₃H₇Cl₃Si
MDL Number:MFCD00018852