Order Entry
Germany
ContactUsLinkComponent
5-Cyano-2-thiopheneboronic acid
5-Cyano-2-thiopheneboronic acid
Catalog # MOLE27885742-1G
Supplier:  Molekula
CAS Number:  
5-Cyano-2-thiopheneboronic acid
Catalog # MOLE27885742-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :152.97 g/mol
CAS Number:305832-67-1
Formula:C₅H₄BNO₂S
MDL Number:MFCD02094029