Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4-(Boc-amino)cyclohexanone

Marketsource
Supplier:   FLUOROCHEM
4-(Boc-amino)cyclohexanone
Supplier:  FLUOROCHEM
Marketsource
Certificates
4-(Boc-amino)cyclohexanone

2 Options Available Below

Product Details & Documents
Synonyms:  4-Aminocyclohexan-1-one, N-BOC protected, tert-Butyl (4-oxocyclohexyl)carbamate
  • CAS Number:179321-49-4
  • Formula:C₁₁H₁₉NO₃
  • MDL Number:MFCD00798168
  • Molecular Weight:213.2700 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource