Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-N-isopropyl-2-nitroaniline
4-Chloro-N-isopropyl-2-nitroaniline
4-Chloro-N-isopropyl-2-nitroaniline
4-Chloro-N-isopropyl-2-nitroaniline
Supplier:
Thermo Scientific
Certificates
Expand image
4-Chloro-N-isopropyl-2-nitroaniline
Supplier:
Thermo Scientific
About this item
CAS Number:
89659-66-5
:
Formula:
C₉H₁₁ClN₂O₂
MDL Number:
MFCD00084958