N(α)-Benzyloxycarbonyl-L-lysine, Sigma-Aldrich®
Catalog # SIAL359793-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:280.3 g/mol
:M-Clarity™ quality level = MQ200
EINECS:218-662-6
CAS Number:2212-75-1
Formula:C₁₄H₂₀N₂O₄
MDL Number:MFCD00038204