Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dibromo-2-(trichloroacetyl)pyrrole
2,3-Dibromo-2-(trichloroacetyl)pyrrole
2,3-Dibromo-2-(trichloroacetyl)pyrrole
2,3-Dibromo-2-(trichloroacetyl)pyrrole
Supplier:
BIONET RESEARCH
Certificates
Expand image
2,3-Dibromo-2-(trichloroacetyl)pyrrole
Supplier:
BIONET RESEARCH
About this item
CAS Number:
50371-52-3
:
Formula:
C₆H₂Br₂Cl₃NO
MDL Number:
MFCD00662082