Order Entry
Germany
ContactUsLinkComponent
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester, Sigma-Aldrich®
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester, Sigma-Aldrich®
Catalog # SIAL683566-1G
Supplier:  Merck
CAS Number:  
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester, Sigma-Aldrich®
Catalog # SIAL683566-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :280.1278 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:864754-16-5
Formula:C₁₃H₂₁BN₂O₄
MDL Number:MFCD08706026