N(α)-Boc-D-tryptophan
Catalog # BACE4003847.0025
Supplier: BACHEM BIOCHEMICA
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:304.3 g/mol
EINECS:226-042-1
CAS Number:5241-64-5
Formula:C₁₆H₂₀N₂O₄
MDL Number:MFCD00037944