Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(Triphenylstannyl)diphenylmethylsilane
(Triphenylstannyl)diphenylmethylsilane
(Triphenylstannyl)diphenylmethylsilane
(Triphenylstannyl)diphenylmethylsilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
(Triphenylstannyl)diphenylmethylsilane
Supplier:
CHEMPUR SILANE
About this item
:
Formula:
C₃₁H₂₈SiSn
MDL Number:
MFCD03093833
:
GHS06
:
GHS09