Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(Triphenylstannyl)dimethylphenylsilane
(Triphenylstannyl)dimethylphenylsilane
(Triphenylstannyl)dimethylphenylsilane
(Triphenylstannyl)dimethylphenylsilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
(Triphenylstannyl)dimethylphenylsilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
210362-76-8
:
Formula:
C₂₆H₂₆SiSn
MDL Number:
MFCD03093830
:
GHS06
:
GHS09