(Tributylstannyl)trimethylsilane
Catalog # SILNSILS16775.5
Supplier: CHEMPUR SILANE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:GHS06
:GHS08
:GHS09
CAS Number:17955-46-3
Formula:C₁₅H₃₆SiSn
MDL Number:MFCD00054903