Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3-Aminopropyl)diisopropylethoxysilane
(3-Aminopropyl)diisopropylethoxysilane
(3-Aminopropyl)diisopropylethoxysilane
(3-Aminopropyl)diisopropylethoxysilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
(3-Aminopropyl)diisopropylethoxysilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
117559-36-1
:
Formula:
C₁₁H₂₇NOSi
MDL Number:
MFCD00270963