(S)-(-)-1-Boc-3-methylpiperazine
Catalog # FLUO011516-5G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:200.28 g/mol
CAS Number:147081-29-6
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD02683204