Order Entry
Germany
ContactUsLinkComponent
Bathocuproin sulphonate disodium salt hydrate for spectrophotometric determination of Cu and Fe, Sigma-Aldrich®
Bathocuproin sulphonate disodium salt hydrate for spectrophotometric determination of Cu and Fe, Sigma-Aldrich®
Certificates
undefined
Bathocuproin sulphonate disodium salt hydrate for spectrophotometric determination of Cu and Fe, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    White to Beige
  • Appearance (Form):
    Powder or Crystals
  • Titration with HClO₄:
    ≥90%
  • Solubility (Colour):
    Colourless to Very Dark Yellow to Very Dark Brown and Colourless to Very Dark Red to Very Dark Purple
  • Solubility (Turbidity), 50 mg/ml in Water:
    Clear to Hazy
  • Water:
    ≤12.0%
  • Infrared Spectrum:
    Conforms to Structure
  • Application Test, Suitable for Use in the Determination of Copper:
    Corresponds to Requirements
  • Assay (ICP):
    7.2 - 9.1%

Specification Test Results