1,4'-Bipiperidine, Sigma-Aldrich®
Catalog # SIAL534498-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:168.28 g/mol
:M-Clarity™ quality level = MQ200
EINECS:225-522-8
CAS Number:4897-50-1
Formula:C₁₀H₂₀N₂
MDL Number:MFCD00006475