0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standard, Dimetridazole 10 µg/ml in methanol
Organic reference standard, Dimetridazole 10 µg/ml in methanol
PROONE9245
:
LGC Standards PROMOCHEM
:
Organic reference standard, Dimetridazole 10 µg/ml in methanol
PROONE9245
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
10 mL
CAS No.:
551-92-8
Description:
Dimetridazole 10 µg/ml in methanol
Formula:
C₅H₇N₃O₂
Description:
Dimetridazole