Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-Methionine sulphoxide
L-Methionine sulphoxide
L-Methionine sulphoxide
L-Methionine sulphoxide
Supplier:
BACHEM BIOCHEMICA
Certificates
Expand image
L-Methionine sulphoxide
Supplier:
BACHEM BIOCHEMICA
About this item
CAS Number:
3226-65-1
:
Formula:
C₅H₁₁NO₃S
MDL Number:
MFCD00063093
:
165.2 g/mol
EINECS:
221-758-0