2-(3-Hydroxypropoxy)benzonitrile
Glass bottle for solids, 1 g
Catalog # ENAMEN300102315.1G
Marketsource
Supplier: SIA ENAMINE
2-(3-Hydroxypropoxy)benzonitrile
Catalog # ENAMEN300102315.1G
Supplier: SIA ENAMINE
Marketsource
CAS Number: 1094453-32-3
Jump to:
Product Details & Documents
- CAS Number:1094453-32-3
- Formula:C₁₀H₁₁NO₂
- MDL Number:MFCD11204586
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



