(±)-1-Boc-3-aminopyrrolidine
Catalog # ENAMEN300-48597.5G
Supplier: SIA ENAMINE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:186.25 g/mol
CAS Number:186550-13-0
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD01861220