Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Anisomycin

Supplier:   Cayman Chemical
Anisomycin
Supplier:  Cayman Chemical
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate
  • CAS Number:22862-76-6
  • Formula:C₁₄H₁₉NO₄
  • MDL Number:MFCD00077650
  • Molecular Weight:265.31 g/mol
  • EINECS:245-269-7
  • UN:3462
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 49,00 €
    49,00 €
    1 * 5 mg
       49,00 €

  • Glass bottle for solids
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 57,00 €
    57,00 €
    1 * 10 mg
       57,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.