Forskolin
Supplier: Cayman Chemical
Jump to:
About this item
Synonyms: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
- CAS Number:66575-29-9
- Formula:C₂₂H₃₄O₇
- MDL Number:MFCD00082317
- Molecular Weight:410.51 g/mol
- EINECS:266-410-9
4 Options Available Below
Product Details & Documents
Synonyms: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
- CAS Number:66575-29-9
- Formula:C₂₂H₃₄O₇
- MDL Number:MFCD00082317
- Molecular Weight:410.51 g/mol
- EINECS:266-410-9
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



