This automated filling system represents a rapid, accurate and cost-effective solution. Consisting of a high precision dispenser PMi and a highly repeatable robotic module XY500, it ensures the automated filling of tubes, bottles, flasks for enhanced laboratory productivity.
PMi Peristaltic Pump
For the automated dispensing of borth or agar, in plates, tubes or bottles.
The PMi automatically dispenses volumes from 1 ml to 20 L with a high degree of accuracy. It has an alpha numeric keyboard which, when combined with a printer, allows full integration within GLP- compliant laboratories.
Accuracy and repetability compliant with ISO 7218
Automatic traceability
Bidirectional connectivity with LiMS communication
Smart software and colour-changing LEDs inform user of system status
RFID-capability
Intuitive interface (help function is with one-click access) with program management
Manual and automatic modes
three adjustable and one customisable speed
XY500 Robotic Module
When used in conjunction with the PMi Peristaltic Pump, the XY500 automates filling in flasks, bottles.
Two racks of 252 tubes each can be filled in the same cycle using dynamic loading directly from the original packaging
4 different 96 tubes rack are available
Easy to program using the menu-driven keypad
Automatic repositioning of the dispensing tip by stepper-motor ensures a greater accuracy and repeatability of filling
The stand-by mode keeps the culture media in suspension
Switch from tube to bottle filling in seconds
Fill over 1000 tubes/hour
When used in conjunction with the PMi Peristaltic Pump, the XY500 automates filling in flasks, bottles.