Order Entry
Germany
ContactUsLinkComponent
 
Biohazard barrier tapes
  LIMP661-022
Biohazard barrier tapes
  LIMP661-022

 

  • Width:
    25,0 mm
  • Colour:
    Black on yellow
  • Description:
    Barrier tape, high risk
  • Seq:
    01
  • Pk:
    1 Roll

 

 

These biohazard barrier tapes are made of vinyl.

Used for packaging and general use as signs.

Length: 66,0 m