Dialysis tubing, VISKING®
Supplier: Serva
Certificates
About this item
MWCO 12 to 14 kD, regenerated cellulose.
- Resistant to chemicals
- Contain low level of heavy metal and sulfide impurities
- Suitable for pH range 2 to 12 and temperatures 4 to 60 °C