Order Entry
Germany
Orders LinkContactUsLinkComponent
Needle Collection Container

Needle Collection Container

1000 ml, Yellow, 70×173 mm
Catalog # 119-4264
Supplier:   DELTALAB
Needle Collection Container
Catalog # 119-4264
Supplier:  DELTALAB
Needle Collection Container

1000 ml, Yellow, 70×173 mm

Price
7,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Needle collection containers are made of high density polyethylene suitable for waste type III.

  • Can be incinerated without release of toxic fumes
  • Printed with two languages (English and Spanish) on the containers

They features two caps, first cap has a mouth which embodies three different devices to remove needles and the other is designed to close the container temporarily or definitively.

Documents

Specifications
  • Ø×H:
    70×173 mm
  • Capacity:
    1000 ml
  • Colour:
    Yellow
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.