ContactUsLinkComponent
Eyewash bottle
Eyewash bottle
Catalog # DELT19384
Supplier:  DELTALAB
Eyewash bottle
Catalog # DELT19384
Supplier:  DELTALAB
Specifications
  • Description:
    Eyewash bottle, empty, 1000 ml
  • Pk:
    1
Specifications
About this item
Made of low density polyethylene, this eyewash bottle is flexible and easy to squeeze.
  • Has an eye cup with snap-on cap
  • Eyecup has a perforated plate to break up the stream providing a gentle spray to the eye
  • Capped air vent valve prevents drawback of contaminated eye wash into the bottle
  • Contaminated solution exits through a drain tube, which rotates in any direction
  • Instructions for use printed in various languages