Accessories
Supplier: Cytiva
About this item
4 Options Available Below
Product Details & Documents
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 18-1111-03
- 18-1111-05
- 18-1114-43
- 18-1111-26
- W×D×H:
- 260×370×100 mm
- 260×370×100 mm
- 260×370×100 mm
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- 18-1111-03Dummy electrode, PTFE, round tip, max flow rate 100 ml/minSpecifications:
More
Specifications
Cat. No.18-1111-03W×D×H260×370×100 mm - 18-1111-05Conductivity flow cell, titanium/CTFE, 14 μl volume, max. flow rate 100 ml/min, 16" outer diameterConductivity flow cell, titanium/CTFE, 14 μl volume, max. flow rate 100 ml/min, 16" outer diameterLog in for availability1 * 1 ST1 810,00 €Specifications:
More
Specifications
Cat. No.18-1111-05W×D×H260×370×100 mm - 18-1114-43Conductivity flow cell, titanium/EPDM/fluoro plastics, 14 μl volume, with M6 connectionConductivity flow cell, titanium/EPDM/fluoro plastics, 14 μl volume, with M6 connectionLog in for availability1 * 1 ST979,00 €Specifications:
More
Specifications
Cat. No.18-1114-43W×D×H - 18-1111-26pH electrode, round tip, with sealed combination double junction, Ag/AgCl reference, electrolyte bridge of 4M KCl saturated with Ag/AgCl, outer electrolyte bridge of 1M KNO 3, annular ceramic reference junction and low profile
pH membranepH electrode, round tip, with sealed combination double junction, Ag/AgCl reference, electrolyte bridge of 4M KCl saturated with Ag/AgCl, outer electrolyte bridge of 1M KNO 3, annular ceramic reference junction and low profile
pH membraneLog in for availability1 * 1 ST1 470,00 €Specifications:More
Specifications
Cat. No.18-1111-26W×D×H260×370×100 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



