Orders LinkContactUsLinkComponent
Replacement plungers
Replacement plungers
Supplier:   Agilent
Replacement plungers
Supplier:  Agilent
Certificates
Replacement plungers
Replacement plungers
7 Options Available Below
Product Details & Documents
Replacement plungers designed for long-lasting applications that demand precision and productivity.
  • Strongly recommended for high-throughput applications
  • X-type syringes feature zero carryover
Product Family Options
Product Information
  • For
    Availability
    Price
  • HEWLG4200-80103
    Replacement plunger
    Replacement plunger
    10 µl syringes
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST
  • HEWLG4200-80105
    Replacement plunger
    Replacement plunger
    100 µl syringes
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST
  • HEWLG4200-80101
    Replacement plunger
    Replacement plunger
    1000 µl syringes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWLG4200-80107
    Replacement plunger
    Replacement plunger
    2500 µl syringes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWLG4200-80108
    Replacement plunger
    Replacement plunger
    5000 µl syringes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWLG4200-80112
    Replacement plunger
    Replacement plunger
    25 µl X-type syringes
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST
  • HEWLG4200-80111
    Replacement plunger
    Replacement plunger
    100 µl X-type syringes
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST