Boiling stones
Catalog # VERND1069103
Supplier: Saint Gobain Life Sciences
Specifications
- Description:PTFE boiling stones
- Pk:450 g
Specifications
About this item
Ultra-pure, virgin PTFE boiling stones promote gentle, efficient boiling and help prevent overheating, even of highly volatile liquids. Ideal for continous extraction methods. Stones minimise 'bumping' during reflux and distillation.
- Chemical resistant
- Ideal for boiling organic liquids
- Minimises superheating
The boiling stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis. They are easily cleaned and non-contaminating. Shows no trace of face acidity or alkalinity. They will not scratch or abrade glassware and leave no residue. Usable up to 288 °C.