Order Entry
Germany
ContactUsLinkComponent
 
Stoppers for 20 mm certified headspace crimp top vials
  548-0774
 :  Agilent
undefined
Stoppers for 20 mm certified headspace crimp top vials
  548-0774
 :  Agilent

 

  • Type:
    Butyl grey, -40 to + 120 °C
  • Seq:
    15
  • Pk:
    100