Orders LinkContactUsLinkComponent
pH electrodes, digital, Edge™
pH electrodes, digital, Edge™
pH electrode HI 12301, gel, with double junction, temperature sensor and Sensor Check**, pH electrode HI 12301, with temp. sensor and Sensor Check™, 0 - 13, Plastic, Gel, Single ceramic, easyPlug 3,5 mm, Field applications
Catalog # 662-1665
Supplier:   Hanna
pH electrodes, digital, Edge™
Catalog # 662-1665
Supplier:  Hanna
pH electrodes, digital, Edge™
pH electrodes, digital, Edge™
pH electrode HI 12301, gel, with double junction, temperature sensor and Sensor Check**, pH electrode HI 12301, with temp. sensor and Sensor Check™, 0 - 13, Plastic, Gel, Single ceramic, easyPlug 3,5 mm, Field applications
Price
284,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Connection:
    easyPlug 3,5 mm
  • For:
    Field applications
  • Type:
    pH electrode HI 12301, with temp. sensor and Sensor Check™
  • Description:
    pH electrode HI 12301, gel, with double junction, temperature sensor and Sensor Check**
  • Material:
    Plastic
  • Diaphr.:
    Single ceramic
  • Electrolyte:
    Gel
  • pH:
    0 - 13