Orders LinkContactUsLinkComponent
Closed caps, PTFE lined
Closed caps, PTFE lined
PTFE faced silicone liner, 24-414, White
Artikelnummer EPSCB267-2414UBB
   Low Manufacturing Impact
Hersteller:   EP SCIENTIFIC
Closed caps, PTFE lined
Artikelnummer EPSCB267-2414UBB
Hersteller:  EP SCIENTIFIC
   Low Manufacturing Impact
Closed caps, PTFE lined
Closed caps, PTFE lined
PTFE faced silicone liner, 24-414, White
Preis
137,00 €
Log in for availability
1 * 144 ST
Product Details & Documents
These PTFE-lined closed caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials.
The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.
Specifications
  • Thread:
    24-414
  • Colour:
    White
  • Description:
    PTFE faced silicone liner
  • Seq:
    01
  • Cat. No.:
    EPSCB267-2414UBB
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact