ContactUsLinkComponent
Thermal cyclers, Mastercycler® pro, individual modules
Thermal cyclers, Mastercycler® pro, individual modules
Catalog # EPPE6320000.007
Supplier:  EPPENDORF
undefined
Thermal cyclers, Mastercycler® pro, individual modules
Catalog # EPPE6320000.007
Supplier:  EPPENDORF
Specifications
  • Description:
    Control panel with connection cable
  • Electrical supply:
    230 V, 50/60 Hz
  • Pk:
    1
Specifications
About this item
Mastercycler pro is an open and flexible system which offers flexibility with fast heating and cooling rates. Systems are available as complete packages or separate modules.
  • Temperature control range 4 to 99 °C
  • Operates in three different configurations
  • Optional self-test dongle available (please enquire for details)
* A Control Panel or CycleManager pro software (both sold separately) is required for operation. CAN-Bus connection cables are required to link cyclers in a network.