Shelving systems for water-endangering and aggressive liquids, KG-GR WPE
Supplier: LACONT UMWELTTECHNIK
Certificates
About this item
Shelving systems are used for the storage of water-endangering and aggressive liquids.
- Galvanised shelving parts and tray-shaped shelves with PE insert
- Height-adjustable shelves at intervals of 25 mm
- Mixture of basic and extension units allows extension as required
- Stable construction by reinforced struts
Two different combinable shelf widths (1000 and 1300 mm) allow optimal utilisation of existing storage space.
Maximum unit load: 1200 kg.