Order Entry
Germany
ContactUsLinkComponent
Dehydrated culture media, TOS-propionate agar (base)
Dehydrated culture media, TOS-propionate agar (base)
Catalog # 1.00043.0500
Supplier:  Merck
CAS Number:  
undefined
Dehydrated culture media, TOS-propionate agar (base)
Catalog # 1.00043.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 g
  • Standards:
    ISO 29981, IDF 220:2010
  • Description:
    TOS-Propionate agar (base)
  • Seq:
    2

Specifications

About this item

In combination with the selective supplement Lithium-Mupirocin (MUP) the TOS Propionate agar allows the direct detection of viable bifidobacteria in starter cultures and various milk products like fermented and unfermented milk, milk powder and baby food. The growth of other lactic acid bacteria is very much inhibited so that a confirmation of presumptive bifodobacteria is in most cases not necessary.

  • The detection is done by using the plate count method with anaerobic incubation at 37 °C