Orders LinkContactUsLinkComponent
Drum stands, FB
Drum stands, FB
Drum stands, FB
Drum stands with fixed drum supports for 60 to 200 L drums, completely galvanised. Tilted support onwards for easy drum emptying.
These drum stands can be combined with compact sump pallets, water protection pallets, hazardous substance stations, floor safety elements and drum carts.
Product Family Options
  • Description
    W×D×H
  • Drum stands, FB 1, fire-galvanised, capacity 1×200 l drum
    600×650×415 mm
    1 * 1 ST
    1 * 1 ST
  • Drum stands, FB 1, fire-galvanised with roller support, capacity 1×200 l drum
    600×650×415 mm
    1 * 1 ST
    1 * 1 ST
  • Drum stands, FB 2, fire-galvanised, capacity 2×200 l drum
    1230×650×415 mm
    1 * 1 ST
    1 * 1 ST
  • Drum stands, FB 2, fire-galvanised with roller support, capacity 2×200 l drum
    1230×650×415 mm
    1 * 1 ST
    1 * 1 ST
  • Drum stands, FB 3, fire-galvanised, capacity 3×60 l drum
    1230×650×415 mm
    1 * 1 ST
    1 * 1 ST
  • Drum stands, FB 3, fire-galvanised with roller support, capacity 3×60 l drum
    1230×650×415 mm
    1 * 1 ST
    1 * 1 ST