Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2-Acetamidophenyl)boronic acid
(2-Acetamidophenyl)boronic acid
(2-Acetamidophenyl)boronic acid
(2-Acetamidophenyl)boronic acid
Supplier:
COMBI-BLOCKS
Certificates
Expand image
(2-Acetamidophenyl)boronic acid
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
169760-16-1
:
Formula:
C₈H₁₀BNO₃
MDL Number:
MFCD02179474
:
178.98 g/mol