Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DL-Methionine methyl ester hydrochloride
DL-Methionine methyl ester hydrochloride
DL-Methionine methyl ester hydrochloride
DL-Methionine methyl ester hydrochloride
ENAMEN30009315.1G
:
SIA ENAMINE
:
DL-Methionine methyl ester hydrochloride
ENAMEN30009315.1G
:
SIA ENAMINE
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
199.7 g/mol
:
240-285-0
:
16118-36-8
:
C₆H₁₃NO₂S·HCl
:
MFCD00067540