
7-Isoquinolinol
Catalog # ENAMEN30005942.1G
Supplier: SIA ENAMINE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:145.16 g/mol
:GHS07
CAS Number:7651-83-4
:14476-74-5
Formula:C₉H₇NO
MDL Number:MFCD00456131