1-(2-Bromoethoxy)-4-methoxybenzene
Catalog # ENAMEN30004497.1G
Supplier: SIA ENAMINE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:231.09 g/mol
CAS Number:22921-76-2
Formula:C₉H₁₁BrO₂
MDL Number:MFCD00017893