Dimethylcyclopropane-1,1-dicarboxylate
Catalog # ENAMEN300100932.1G
Supplier: SIA ENAMINE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:158.15 g/mol
EINECS:414-240-2
CAS Number:6914-71-2
Formula:C₇H₁₀O₄
MDL Number:MFCD00192079