
L(+)-Lactic acid ≥90% in aqueous solution, extra pure
:79-33-4
:
:C₃H₆O₃
:MFCD00064266
:90.08 g/mol
:1.21 g/cm³ (20 °C)
:201-196-2
:GHS05
:Refrigerator
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals