6-Bromoquinaldine
Glass bottle for solids, 1 g
Catalog # ENAMEN300104966.1G
Marketsource
Supplier: SIA ENAMINE
6-Bromoquinaldine
Catalog # ENAMEN300104966.1G
Supplier: SIA ENAMINE
Marketsource
CAS Number: 877-42-9
Jump to:
Product Details & Documents
- CAS Number:877-42-9
- Formula:C₁₀H₈BrN
- MDL Number:MFCD00079724
- Molecular Weight:222.08 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



