2-Deoxy-D(-)-ribose
Glass bottle for solids, 1 g
ENAMEN300106817.1G
Marketsource
: 2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal
- :533-67-5
- :C₅H₁₀O₄
- :MFCD00135904
- :134.13 g/mol
- :13,02925
- :292 °C (1013 hPa)
- :95…97 °C
- :Ambient
- :208-573-0
- Pack type:Glass bottle for solids
- Pk:1 g