2-(Trifluoromethyl)pyridine
Glass bottle for solids, 1 g
Catalog # ENAMEN300104803.1G
Marketsource
Supplier: SIA ENAMINE
2-(Trifluoromethyl)pyridine
Catalog # ENAMEN300104803.1G
Supplier: SIA ENAMINE
Marketsource
CAS Number: 368-48-9
Jump to:
Product Details & Documents
- CAS Number:368-48-9
- Formula:C₆H₄F₃N
- MDL Number:MFCD02183562
- Molecular Weight:147.1 g/mol
- Boiling Pt:138…139 °C (1013 hPa)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



