
1-(4-Chlorophenyl)-2-thiourea
Catalog # ENAMEN30006963.1G
Supplier: SIA ENAMINE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:304 °C (1013 hPa)
Melting Pt:201…201.5 °C
UN:2811
ADR:6.1,II
:186.66 g/mol
EINECS:223-022-4
:GHS06
CAS Number:3696-23-9
Formula:(C₆H₄Cl)NC(S)NH₂
MDL Number:MFCD00022168