Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Mercapto-1,2-propanediol
3-Mercapto-1,2-propanediol
3-Mercapto-1,2-propanediol
ENAMEN300100297.1G
:
SIA ENAMINE
:
3-Mercapto-1,2-propanediol
ENAMEN300100297.1G
:
SIA ENAMINE
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
108.16 g/mol
:
12,09471
:
118 °C (1013 hPa)
:
1.247 g/cm³ (20 °C)
:
202-495-0
:
GHS07
:
Ambient
:
96-27-5
:
C₃H₈O₂S
:
MFCD00004879