Order Entry
Germany
ContactUsLinkComponent
Titanium(IV) oxide 99.0-100.5%, AnalaR NORMAPUR®
Titanium(IV) oxide 99.0-100.5%, AnalaR NORMAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Titanium(IV) oxide 99.0-100.5%, AnalaR NORMAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    99.0 - 100.5 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Loss on drying (105°C):
    Max. 0.5 %
  • Loss on ignition (800°C):
    Max. 0.5 %
  • Solubility in hot water:
    Max. 0.5 %
  • Solubility in hydrochloric acid:
    Max. 0.5 %
  • As (Arsenic):
    Max. 1 ppm
  • Fe (Iron):
    Max. 0.02 %
  • Sb (Antimony):
    Max. 100 ppm

Specification Test Results

Related Information